siRNA Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040715.1 | NP_001035805.1 | 7 | 2604 | ||
GenBank mRNA | AK092303.1 | - | 3 | 2030 | ||
AK094787.1 | - | 8 | 1941 | |||
AK096855.1 | - | 6 | 1300 | |||
AL834156.1 | - | 8 | 2708 | |||
BC007594.1 | - | 6 | 1798 | |||
BC128539.1 | - | 5 | 1144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
