siRNA Details
Gene Symbol: AGRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001138.1 | NP_001129.1 | 4 | 649 | ||
GenBank mRNA | BC110443.2 | - | 3 | 352 | 40002498 | |
U88063.1 | - | 4 | 649 | |||
U89485.1 | - | 3 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
