siRNA Details
Gene Symbol: AGRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001138.1 | NP_001129.1 | 2 | 400 | ||
GenBank mRNA | BC110443.2 | - | 1 | 103 | 40002498 | |
U88063.1 | - | 2 | 400 | |||
U89485.1 | - | 1 | 100 |

More Information
Other Species Targeted

Gene Ontology Categories:
