siRNA Details
Gene Symbol: ZFYVE28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172656.1 | NP_001166127.1 | 3 | 531 | ||
NM_001172657.1 | NP_001166128.1 | 3 | 531 | |||
NM_001172658.1 | NP_001166129.1 | 2 | 390 | |||
NM_001172659.1 | NP_001166130.1 | 3 | 564 | |||
NM_001172660.1 | NP_001166131.1 | 3 | 564 | |||
NM_020972.2 | NP_066023.2 | 3 | 531 | |||
XM_006713900.3 | XP_006713963.1 | 3 | 585 | |||
XM_006713902.3 | XP_006713965.1 | 3 | 586 | |||
XM_006713904.3 | XP_006713967.1 | 3 | 585 | |||
XM_011513541.2 | XP_011511843.1 | 3 | 585 | |||
XM_017008502.1 | XP_016863991.1 | 3 | 307 | |||
XM_017008503.1 | XP_016863992.1 | 3 | 586 | |||
XM_017008504.1 | XP_016863993.1 | 3 | 585 | |||
XM_017008505.1 | XP_016863994.1 | 3 | 307 | |||
XM_017008506.1 | XP_016863995.1 | 3 | 586 | |||
XM_017008507.1 | XP_016863996.1 | 3 | 585 | |||
XM_017008508.1 | XP_016863997.1 | 3 | 585 | |||
XM_017008509.1 | XP_016863998.1 | 3 | 585 | |||
XM_017008510.1 | XP_016863999.1 | 3 | 585 | |||
GenBank mRNA | AB046863.1 | - | 3 | 530 | ||
AK293775.1 | - | 1 | 14 | |||
AK294710.1 | - | 4 | 358 | |||
AK316484.1 | - | 3 | 564 | |||
BC053664.1 | - | 3 | 497 | |||
BC137309.1 | - | 4 | 274 | 9021306 | ||
BC137310.1 | - | 4 | 274 | 9021307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
