siRNA Details
Gene Symbol: ZFYVE28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172656.1 | NP_001166127.1 | 1 | 339 | ||
NM_001172657.1 | NP_001166128.1 | 1 | 339 | |||
NM_001172658.1 | NP_001166129.1 | 1 | 339 | |||
NM_020972.2 | NP_066023.2 | 1 | 339 | |||
XM_006713900.3 | XP_006713963.1 | 1 | 393 | |||
XM_006713902.3 | XP_006713965.1 | 1 | 394 | |||
XM_006713904.3 | XP_006713967.1 | 1 | 393 | |||
XM_011513541.2 | XP_011511843.1 | 1 | 393 | |||
XM_017008503.1 | XP_016863992.1 | 1 | 394 | |||
XM_017008504.1 | XP_016863993.1 | 1 | 393 | |||
XM_017008506.1 | XP_016863995.1 | 1 | 394 | |||
XM_017008507.1 | XP_016863996.1 | 1 | 393 | |||
XM_017008508.1 | XP_016863997.1 | 1 | 393 | |||
XM_017008509.1 | XP_016863998.1 | 1 | 393 | |||
XM_017008510.1 | XP_016863999.1 | 1 | 393 | |||
GenBank mRNA | AB046863.1 | - | 1 | 338 | ||
AK294710.1 | - | 2 | 166 | |||
BC053664.1 | - | 1 | 305 | |||
BC137309.1 | - | 2 | 82 | 9021306 | ||
BC137310.1 | - | 2 | 82 | 9021307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
