siRNA Details
Gene Symbol: CALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005184.2 | NP_005175.2 | 6 | 1886 | ||
GenBank mRNA | AK094964.1 | - | 3 | 2165 | ||
AL050207.1 | - | 2 | 498 | |||
BC005137.2 | - | 6 | 1790 | 3504793 | ||
BC006182.1 | - | 6 | 1718 | 3528814 | ||
FJ224310.1 | - | 6 | 1803 | |||
J04046.1 | - | 6 | 1803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
