siRNA Details
Gene Symbol: WDR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260474.1 | NP_001247403.1 | 4 | 415 | ||
NM_001260475.1 | NP_001247404.1 | 4 | 363 | |||
NM_001260476.1 | NP_001247405.1 | 4 | 263 | |||
NM_001260477.1 | NP_001247406.1 | 4 | 470 | |||
NM_018669.5 | NP_061139.2 | 4 | 415 | |||
NM_033661.4 | NP_387510.1 | 4 | 415 | |||
NR_048535.1 | 4 | 307 | ||||
XM_011529433.1 | XP_011527735.1 | 4 | 678 | |||
GenBank mRNA | AJ243912.1 | - | 4 | 359 | ||
AJ243913.1 | - | 4 | 359 | |||
AK056343.1 | - | 4 | 470 | |||
AK292923.1 | - | 4 | 356 | |||
AK298015.1 | - | 4 | 307 | |||
AK315008.1 | - | 4 | 306 | |||
BC001074.2 | - | 4 | 242 | 2821060 | ||
BC006341.2 | - | 4 | 363 | 4080041 | ||
BG722541.1 | - | 4 | 389 | |||
BU541429.1 | - | 4 | 341 | |||
DC303088.1 | - | 4 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
