siRNA Details
Gene Symbol: WDR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260474.1 | NP_001247403.1 | 11 | 1176 | ||
NM_001260475.1 | NP_001247404.1 | 11 | 1127 | |||
NM_001260476.1 | NP_001247405.1 | 11 | 1027 | |||
NM_001260477.1 | NP_001247406.1 | 11 | 1234 | |||
NM_018669.5 | NP_061139.2 | 11 | 1179 | |||
NM_033661.4 | NP_387510.1 | 11 | 1179 | |||
NR_048535.1 | 11 | 1071 | ||||
XM_011529433.1 | XP_011527735.1 | 11 | 1442 | |||
XM_017028262.1 | XP_016883751.1 | 9 | 1077 | |||
XM_017028263.1 | XP_016883752.1 | 9 | 836 | |||
XM_017028264.1 | XP_016883753.1 | 9 | 836 | |||
GenBank mRNA | AJ243912.1 | - | 12 | 1123 | ||
AJ243913.1 | - | 12 | 1123 | |||
AK056343.1 | - | 11 | 1234 | |||
AK292923.1 | - | 11 | 1117 | |||
AK298015.1 | - | 11 | 1071 | |||
AK315008.1 | - | 11 | 1070 | |||
BC001074.2 | - | 11 | 1006 | 2821060 | ||
BC006341.2 | - | 11 | 1127 | 4080041 | ||
BC018866.2 | - | 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
