siRNA Details
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286258.1 | NP_001273187.1 | 10 | 2496 | ||
NM_001320694.1 | NP_001307623.1 | 11 | 2665 | |||
NM_004571.4 | NP_004562.2 | 11 | 2668 | |||
GenBank mRNA | AY196965.1 | - | 11 | 2646 | ||
U68727.1 | - | 10 | 2530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
