siRNA Details
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286258.1 | NP_001273187.1 | 10 | 1504 | ||
NM_001320694.1 | NP_001307623.1 | 11 | 1673 | |||
NM_004571.4 | NP_004562.2 | 11 | 1676 | |||
GenBank mRNA | AK294794.1 | - | 10 | 1373 | ||
AY196965.1 | - | 11 | 1654 | |||
BC007746.2 | - | 10 | 1483 | 4099156 | ||
U68727.1 | - | 10 | 1547 | |||
Y13613.1 | - | 11 | 1589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
