siRNA Details
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286258.1 | NP_001273187.1 | 7 | 849 | ||
NM_001320694.1 | NP_001307623.1 | 8 | 1018 | |||
NM_004571.4 | NP_004562.2 | 8 | 1021 | |||
GenBank mRNA | AK222892.1 | - | 7 | 887 | ||
AK294794.1 | - | 7 | 718 | |||
AY142115.1 | - | 8 | 998 | |||
AY196965.1 | - | 8 | 999 | |||
BC000735.2 | - | 8 | 967 | |||
BC007746.2 | - | 7 | 828 | 4099156 | ||
U68727.1 | - | 7 | 892 | |||
Y13613.1 | - | 8 | 934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
