siRNA Details
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 17 | 2796 | ||
XM_011529714.2 | XP_011528016.1 | 18 | 2900 | |||
XM_011529716.2 | XP_011528018.1 | 17 | 2753 | |||
XM_011529717.2 | XP_011528019.1 | 18 | 2900 | |||
XM_011529718.2 | XP_011528020.1 | 18 | 2900 | |||
XM_011529719.2 | XP_011528021.1 | 16 | 2456 | |||
XM_011529720.2 | XP_011528022.1 | 18 | 2900 | |||
XM_011529721.2 | XP_011528023.1 | 15 | 2273 | |||
XM_011529722.2 | XP_011528024.1 | 15 | 2239 | |||
XM_011529723.2 | XP_011528025.1 | 14 | 2136 | |||
XM_011529725.2 | XP_011528027.1 | 18 | 2900 | |||
XM_011529726.2 | XP_011528028.1 | 18 | 2900 | |||
XM_011529727.2 | XP_011528029.1 | 14 | 2141 | |||
XM_011529728.2 | XP_011528030.1 | 15 | 2422 | |||
XM_011529729.2 | XP_011528031.1 | 14 | 2046 | |||
XM_011529730.2 | XP_011528032.1 | 18 | 2895 | |||
XM_017028454.1 | XP_016883943.1 | 16 | 2642 | |||
XM_017028455.1 | XP_016883944.1 | 10 | 1642 | |||
GenBank mRNA | BC101728.1 | - | 17 | 2716 | 8069234 | |
U19252.1 | - | 17 | 2764 | |||
U61500.1 | - | 17 | 2780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
