siRNA Details
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 19 | 3144 | ||
XM_005261168.4 | XP_005261225.1 | 19 | 3014 | |||
XM_011529714.2 | XP_011528016.1 | 20 | 3248 | |||
XM_011529715.2 | XP_011528017.1 | 20 | 3125 | |||
XM_011529716.2 | XP_011528018.1 | 19 | 3101 | |||
XM_011529717.2 | XP_011528019.1 | 20 | 3248 | |||
XM_011529718.2 | XP_011528020.1 | 20 | 3248 | |||
XM_011529719.2 | XP_011528021.1 | 18 | 2804 | |||
XM_011529720.2 | XP_011528022.1 | 20 | 3248 | |||
XM_011529721.2 | XP_011528023.1 | 17 | 2621 | |||
XM_011529722.2 | XP_011528024.1 | 17 | 2587 | |||
XM_011529723.2 | XP_011528025.1 | 16 | 2484 | |||
XM_011529727.2 | XP_011528029.1 | 16 | 2489 | |||
XM_011529728.2 | XP_011528030.1 | 17 | 2770 | |||
XM_011529729.2 | XP_011528031.1 | 16 | 2394 | |||
XM_017028454.1 | XP_016883943.1 | 18 | 2990 | |||
XM_017028455.1 | XP_016883944.1 | 12 | 1990 | |||
GenBank mRNA | AK295369.1 | - | 6 | 764 | ||
AK300912.1 | - | 3 | 302 | |||
AK303570.1 | - | 7 | 749 | |||
AL833668.1 | - | 2 | 2000 | |||
BC101728.1 | - | 19 | 3064 | 8069234 | ||
U19252.1 | - | 19 | 3112 | |||
U61500.1 | - | 19 | 3128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
