siRNA Details
Gene Symbol: SIGLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033329.2 | NP_201586.1 | 7 | 1782 | ||
NM_053003.3 | NP_443729.1 | 8 | 2172 | |||
GenBank mRNA | AY358140.1 | - | 8 | 2130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
