siRNA Details
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 3 | 169 | ||
NM_021603.3 | NP_067614.1 | 3 | 176 | |||
GenBank mRNA | AA309757.1 | - | 3 | 164 | ||
AF241236.1 | - | 3 | 144 | |||
AK096069.1 | - | 2 | 367 | |||
BC005302.1 | - | 3 | 176 | 3828677 | ||
BC013289.1 | - | 3 | 172 | 3686762 | ||
BT006721.1 | - | 3 | 97 | |||
DQ786191.1 | - | 2 | 322 | |||
U50743.1 | - | 3 | 133 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 8 | 782 | ||
GenBank mRNA | FY211982.1 | - | 8 | 379 | ||
FY214031.1 | - | 8 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
