siRNA Details
Gene Symbol: CUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202543.1 | NP_001189472.1 | 4 | 393 | ||
NM_001202544.2 | NP_001189473.1 | 3 | 345 | |||
NM_001202545.2 | NP_001189474.1 | 4 | 393 | |||
NM_001202546.2 | NP_001189475.1 | 3 | 178 | |||
NM_001913.4 | NP_001904.2 | 4 | 393 | |||
NM_181500.3 | NP_852477.1 | 4 | 393 | |||
NM_181552.3 | NP_853530.2 | 4 | 271 | |||
XM_005250150.2 | XP_005250207.1 | 4 | 792 | |||
XM_005250151.2 | XP_005250208.1 | 4 | 795 | |||
XM_005250154.3 | XP_005250211.1 | 4 | 649 | |||
XM_006715854.2 | XP_006715917.1 | 4 | 792 | |||
XM_006715855.1 | XP_006715918.1 | 4 | 649 | |||
XM_011515823.2 | XP_011514125.1 | 4 | 790 | |||
XM_011515825.1 | XP_011514127.1 | 4 | 358 | |||
XM_017011759.1 | XP_016867248.1 | 4 | 358 | |||
XM_017011760.1 | XP_016867249.1 | 4 | 358 | |||
GenBank mRNA | AB075522.1 | - | 1 | 45 | ||
AK122726.1 | - | 4 | 276 | |||
AK125097.1 | - | 3 | 240 | |||
AK222832.1 | - | 4 | 289 | |||
AK297548.1 | - | 4 | 279 | |||
AK303151.1 | - | 3 | 182 | |||
BC025422.2 | - | 4 | 274 | |||
BC066592.1 | - | 4 | 286 | 5740343 | ||
L12579.1 | - | 4 | 285 | |||
M74099.1 | - | 4 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
