siRNA Details
Gene Symbol: CUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202543.1 | NP_001189472.1 | 2, 3 | 296 | ||
NM_001202545.2 | NP_001189474.1 | 2, 3 | 296 | |||
NM_001913.4 | NP_001904.2 | 2, 3 | 296 | |||
NM_181500.3 | NP_852477.1 | 2, 3 | 296 | |||
NM_181552.3 | NP_853530.2 | 2, 3 | 174 | |||
XM_005250150.2 | XP_005250207.1 | 2, 3 | 695 | |||
XM_005250151.2 | XP_005250208.1 | 2, 3 | 698 | |||
XM_005250154.3 | XP_005250211.1 | 2, 3 | 552 | |||
XM_006715854.2 | XP_006715917.1 | 2, 3 | 695 | |||
XM_006715855.1 | XP_006715918.1 | 2, 3 | 552 | |||
XM_011515823.2 | XP_011514125.1 | 2, 3 | 693 | |||
XM_011515825.1 | XP_011514127.1 | 2, 3 | 261 | |||
XM_017011759.1 | XP_016867248.1 | 2, 3 | 261 | |||
XM_017011760.1 | XP_016867249.1 | 2, 3 | 261 | |||
GenBank mRNA | AK122726.1 | - | 2, 3 | 179 | ||
AK222832.1 | - | 2, 3 | 192 | |||
AK297548.1 | - | 2, 3 | 182 | |||
BC025422.2 | - | 2, 3 | 177 | |||
BC066592.1 | - | 2, 3 | 189 | 5740343 | ||
L12579.1 | - | 2, 3 | 188 | |||
M74099.1 | - | 2, 3 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
