siRNA Details
Gene Symbol: ZNF585A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288800.1 | NP_001275729.1 | 5 | 590 | ||
NM_152655.3 | NP_689868.1 | 6 | 692 | |||
NM_199126.2 | NP_954577.1 | 6 | 632 | |||
GenBank mRNA | AK056389.1 | - | 6 | 639 | ||
AK074345.1 | - | 3 | 224 | |||
AK290660.1 | - | 5 | 569 | |||
BC052956.1 | - | 6 | 632 | |||
BC063820.1 | - | 5 | 524 | 6500797 | ||
DA738522.1 | - | 6 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
