siRNA Details
Gene Symbol: FBXO17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024907.6 | NP_079183.4 | 6 | 1698 | ||
NM_148169.2 | NP_680474.1 | 6 | 1585 | |||
NR_104026.1 | 6 | 1736 | ||||
GenBank mRNA | AF386743.1 | - | 6 | 1628 | ||
BM711851.1 | - | 1 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
