siRNA Details
Gene Symbol: RASGRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146202.1 | NP_001139674.1 | 16 | 2045 | ||
NM_001146203.1 | NP_001139675.1 | 15 | 1811 | |||
NM_001146204.1 | NP_001139676.1 | 16 | 1880 | |||
NM_001146205.1 | NP_001139677.1 | 16 | 1985 | |||
NM_001146206.1 | NP_001139678.1 | 13 | 1520 | |||
NM_001146207.1 | NP_001139679.1 | 14 | 1796 | |||
NM_170604.2 | NP_733749.1 | 16 | 2087 | |||
GenBank mRNA | AF448437.1 | - | 16 | 1944 | ||
AK097292.1 | - | 8 | 1158 | |||
AY048119.1 | - | 16 | 2087 | |||
AY048120.1 | - | 15 | 2202 | |||
AY048121.1 | - | 16 | 2045 | |||
BC146669.1 | - | 16 | 1927 | 40148170 | ||
BC150202.1 | - | 16 | 1927 | 40148808 | ||
FJ768677.1 | - | 15 | 1811 | |||
FJ768678.1 | - | 16 | 1880 | |||
FJ768679.1 | - | 16 | 1985 | |||
FJ768680.1 | - | 13 | 1520 | |||
FJ768681.1 | - | 15 | 2182 | |||
FJ768682.1 | - | 14 | 1796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
