siRNA Details
Gene Symbol: CCL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005064.5 | NP_005055.3 | 3 | 244 | ||
GenBank mRNA | BC143310.1 | - | 3 | 238 | 9051815 | |
U58913.1 | - | 3 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
