siRNA Details
Gene Symbol: CCL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005064.5 | NP_005055.3 | 3 | 269 | ||
NM_145898.3 | NP_665905.2 | 2, 3 | 218 | |||
GenBank mRNA | BC093970.1 | - | 2, 3 | 212 | 7939815 | |
BC143310.1 | - | 3 | 263 | 9051815 | ||
U58913.1 | - | 3 | 269 | |||
U67128.1 | - | 2, 3 | 162 | |||
U85767.1 | - | 2, 3 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
