siRNA Details
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152344.3 | NP_689557.1 | 6 | 1808 | ||
GenBank mRNA | AK055218.1 | - | 5 | 1537 | ||
AL832076.1 | - | 1 | 1496 | |||
BC016791.2 | - | 1 | 1302 | |||
BC044587.1 | - | 5 | 1538 | 4794614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
