siRNA Details
Gene Symbol: KRT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002282.3 | NP_002273.3 | 9 | 1663 | ||
GenBank mRNA | BC126935.1 | - | 9 | 1606 | 40018026 | |
BC128063.1 | - | 9 | 1622 | 40006347 | ||
X99141.1 | - | 1652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
