siRNA Details
Gene Symbol: NEBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096540.1 | - | 2655 |
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936824.1 | - | 6 | 4072 |
Gene Symbol: RPL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000987.3 | NP_000978.1 | 4 | 555 | ||
NM_001315530.1 | NP_001302459.1 | 4 | 848 | |||
NM_001315531.1 | NP_001302460.1 | 4 | 767 | |||
GenBank mRNA | BC071664.1 | - | 4 | 490 | 5189436 | |
BI091167.1 | - | 4 | 555 | |||
BM928617.1 | - | 3 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
