siRNA Details
Gene Symbol: HS3ST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009606.3 | NP_001009606.3 | 2 | 698 | ||
XM_011522608.2 | XP_011520910.1 | 2 | 924 | |||
XM_011522609.2 | XP_011520911.1 | 2 | 2405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
