siRNA Details
Gene Symbol: CABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318496.1 | NP_001305425.1 | 6 | 711 | ||
NM_016366.2 | NP_057450.2 | 6 | 707 | |||
GenBank mRNA | AB593136.1 | - | 6 | 897 | ||
AF169154.1 | - | 6 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
