siRNA Details
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006624.1 | NP_001006625.1 | 6 | 906 | ||
NM_001006625.1 | NP_001006626.1 | 6 | 900 | |||
NM_006474.4 | NP_006465.3 | 6 | 1082 | |||
NM_198389.2 | NP_938203.2 | 6 | 1076 | |||
XM_006710295.1 | XP_006710358.1 | 6 | 813 | |||
GenBank mRNA | AF390106.1 | - | 6 | 988 | ||
AK075327.1 | - | 6 | 1001 | |||
AK307706.1 | - | 7 | 968 | |||
AY194238.1 | - | 6 | 1008 | |||
BC014668.1 | - | 6 | 1038 | 4876446 | ||
BC022812.1 | - | 6 | 1038 | 4875444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
