siRNA Details
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006624.1 | NP_001006625.1 | 6 | 1178 | ||
NM_001006625.1 | NP_001006626.1 | 6 | 1172 | |||
NM_006474.4 | NP_006465.3 | 6 | 1354 | |||
NM_198389.2 | NP_938203.2 | 6 | 1348 | |||
XM_006710295.1 | XP_006710358.1 | 6 | 1085 | |||
GenBank mRNA | AK075345.1 | - | 6 | 1272 | ||
AK307705.1 | - | 6 | 1084 | |||
AK307706.1 | - | 7 | 1240 | |||
AY194238.1 | - | 6 | 1280 | |||
CD245679.1 | - | 1 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
