siRNA Details
Gene Symbol: CPSF3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256456.1 | NP_001243385.1 | 11 | 1435 | ||
NM_001256460.1 | NP_001243389.1 | 10 | 1069 | |||
NM_001256462.1 | NP_001243391.1 | 7 | 668 | |||
NM_001256463.1 | NP_001243392.1 | 7 | 659 | |||
NM_017871.5 | NP_060341.2 | 9 | 962 | |||
XM_011541647.1 | XP_011539949.1 | 11 | 1113 | |||
XM_011541648.1 | XP_011539950.1 | 11 | 1117 | |||
XM_011541650.1 | XP_011539952.1 | 9 | 842 | |||
XM_017001557.1 | XP_016857046.1 | 10 | 918 | |||
XM_017001558.1 | XP_016857047.1 | 11 | 972 | |||
GenBank mRNA | AK000549.1 | - | 1 | 106 | ||
AK021939.1 | - | 7 | 625 | |||
AK023356.1 | - | 9 | 880 | |||
AK056652.1 | - | 11 | 1357 | |||
AK058018.1 | - | 7 | 634 | |||
AK291387.1 | - | 11 | 1398 | |||
AK297350.1 | - | 10 | 1019 | |||
BC013904.2 | - | 5 | 371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
