siRNA Details
Gene Symbol: CPSF3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256456.1 | NP_001243385.1 | 19 | 2534 | ||
NM_001256460.1 | NP_001243389.1 | 18 | 2168 | |||
NM_001256462.1 | NP_001243391.1 | 15 | 1767 | |||
NM_001256463.1 | NP_001243392.1 | 15 | 1758 | |||
NM_017871.5 | NP_060341.2 | 17 | 2061 | |||
XM_011541647.1 | XP_011539949.1 | 19 | 2212 | |||
XM_011541648.1 | XP_011539950.1 | 19 | 2216 | |||
XM_011541650.1 | XP_011539952.1 | 17 | 1941 | |||
XM_017001557.1 | XP_016857046.1 | 18 | 2017 | |||
XM_017001558.1 | XP_016857047.1 | 19 | 2071 | |||
GenBank mRNA | AK000549.1 | - | 9 | 1204 | ||
AK021939.1 | - | 15 | 1724 | |||
AK023356.1 | - | 18 | 1910 | |||
AK056652.1 | - | 19 | 2456 | |||
AK058018.1 | - | 15 | 1733 | |||
AK092190.1 | - | 14 | 1794 | |||
AK123641.1 | - | 6 | 3005 | |||
AK291387.1 | - | 19 | 2497 | |||
AL136813.1 | - | 17 | 2040 | |||
BC007978.2 | - | 17 | 2023 | 3836956 | ||
BC008041.2 | - | 12 | 1385 | 3831587 | ||
BC013904.2 | - | 13 | 1470 | |||
BC020199.1 | - | 12 | 1385 | 4642543 | ||
KF274554.1 | - | 3 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
