siRNA Details
Gene Symbol: CPSF3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256456.1 | NP_001243385.1 | 11 | 1331 | ||
NM_001256460.1 | NP_001243389.1 | 10 | 965 | |||
NM_001256462.1 | NP_001243391.1 | 7 | 564 | |||
NM_001256463.1 | NP_001243392.1 | 7 | 555 | |||
NM_017871.5 | NP_060341.2 | 9 | 858 | |||
XM_011541647.1 | XP_011539949.1 | 11 | 1009 | |||
XM_011541648.1 | XP_011539950.1 | 11 | 1013 | |||
XM_011541650.1 | XP_011539952.1 | 9 | 738 | |||
XM_017001557.1 | XP_016857046.1 | 10 | 814 | |||
XM_017001558.1 | XP_016857047.1 | 11 | 868 | |||
GenBank mRNA | AK021939.1 | - | 7 | 521 | ||
AK023356.1 | - | 9 | 776 | |||
AK056652.1 | - | 11 | 1253 | |||
AK058018.1 | - | 7 | 530 | |||
AK092190.1 | - | 6 | 591 | |||
AK123641.1 | - | 1 | 1261 | |||
AK291387.1 | - | 11 | 1294 | |||
AK297350.1 | - | 10 | 915 | |||
AL136813.1 | - | 9 | 837 | |||
BC000675.2 | - | 6 | 398 | |||
BC007978.2 | - | 9 | 820 | 3836956 | ||
BC008041.2 | - | 6 | 455 | 3831587 | ||
BC013904.2 | - | 5 | 267 | |||
BC020199.1 | - | 6 | 455 | 4642543 | ||
CR533557.1 | - | 9 | 779 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028020, BC011155, AK010425, AK150436, AK172533, and others.
Gene Ontology Categories:
