siRNA Details
Gene Symbol: MEGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001409.3 | NP_001400.3 | 9 | 1219 | ||
XM_006710406.3 | XP_006710469.1 | 6 | 854 | |||
XM_011540885.1 | XP_011539187.1 | 10 | 1524 | |||
XM_011540886.1 | XP_011539188.1 | 10 | 1524 | |||
XM_011540887.2 | XP_011539189.1 | 5 | 732 | |||
XM_011540888.2 | XP_011539190.1 | 10 | 1524 | |||
XM_017000533.1 | XP_016856022.1 | 10 | 1524 | |||
GenBank mRNA | AB011539.2 | - | 6 | 880 | ||
AB231860.1 | - | 9 | 1219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
