siRNA Details
Gene Symbol: MEGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001409.3 | NP_001400.3 | 32 | 4288 | ||
XM_006710406.3 | XP_006710469.1 | 29 | 3923 | |||
XM_011540885.1 | XP_011539187.1 | 33 | 4593 | |||
XM_011540886.1 | XP_011539188.1 | 33 | 4593 | |||
XM_011540887.2 | XP_011539189.1 | 28 | 3801 | |||
XM_017000533.1 | XP_016856022.1 | 32 | 4461 | |||
GenBank mRNA | AB011539.2 | - | 26 | 3559 | ||
AB231860.1 | - | 32 | 4288 | |||
BU501408.1 | - | 7 | 736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
