siRNA Details
Gene Symbol: NTNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000686.1 | XP_016856175.1 | 5 | 3863 | ||
GenBank mRNA | AB023193.1 | - | 5 | 3895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
