siRNA Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 3 | 943 | ||
NM_152366.4 | NP_689579.3 | 3 | 924 | |||
NR_033385.1 | 4 | 693 | ||||
NR_033386.1 | 4 | 712 | ||||
GenBank mRNA | AK093268.1 | - | 1 | 1288 | ||
AY601914.1 | - | 3 | 942 | |||
BC022077.1 | - | 3 | 923 | 4822843 | ||
BC032482.1 | - | 1 | 1317 | 5222141 | ||
BC066896.1 | - | 3 | 918 | 4828667 | ||
BI549017.1 | - | 3 | 536 | |||
BU173744.1 | - | 4 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
