siRNA Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007255.2 | NP_001007256.1 | 3, 4 | 1047 | ||
NM_152366.4 | NP_689579.3 | 3, 4 | 1028 | |||
NR_033385.1 | 4, 5 | 797 | ||||
NR_033386.1 | 4, 5 | 816 | ||||
GenBank mRNA | AY601914.1 | - | 3, 4 | 1046 | ||
BC022077.1 | - | 3, 4 | 1027 | 4822843 | ||
BC066896.1 | - | 3, 4 | 1022 | 4828667 | ||
BU173744.1 | - | 4, 5 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
