siRNA Details
Gene Symbol: MT1HL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276687.1 | NP_001263616.1 | 1 | 85 | ||
GenBank mRNA | AF333388.1 | - | 1 | 85 | ||
BC133030.1 | - | 1 | 79 | |||
BC133032.1 | - | 1 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
