siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 1 | 348 | ||
XM_006711816.3 | XP_006711879.1 | 1 | 170 | |||
XM_017002391.1 | XP_016857880.1 | 1 | 33 | |||
GenBank mRNA | CT004972.1 | - | 1 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
