siRNA Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287584.1 | NP_001274513.1 | 8 | 914 | ||
NM_001287585.1 | NP_001274514.1 | 8 | 934 | |||
NM_138285.4 | NP_612142.2 | 7 | 811 | |||
NR_109856.1 | 6 | 669 | ||||
XM_006712254.2 | XP_006712317.1 | 7 | 776 | |||
XM_011510576.2 | XP_011508878.1 | 9 | 1042 | |||
XM_011510577.2 | XP_011508879.1 | 9 | 1229 | |||
XM_017003308.1 | XP_016858797.1 | 7 | 967 | |||
GenBank mRNA | AF411516.1 | - | 7 | 811 | ||
AF514993.1 | - | 7 | 768 | |||
AK289601.1 | - | 6 | 612 | |||
AK290760.1 | - | 7 | 723 | |||
AK298199.1 | - | 8 | 844 | |||
AK302350.1 | - | 8 | 883 | |||
BC035597.1 | - | 7 | 735 | |||
BC047029.1 | - | 7 | 798 | 5541023 | ||
BC061698.1 | - | 7 | 765 | 4132440 | ||
BI821231.1 | - | 6 | 605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
