siRNA Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287584.1 | NP_001274513.1 | 3 | 364 | ||
NM_001287585.1 | NP_001274514.1 | 2 | 261 | |||
NM_138285.4 | NP_612142.2 | 2 | 261 | |||
NR_109856.1 | 2 | 261 | ||||
XM_006712254.2 | XP_006712317.1 | 2 | 226 | |||
XM_011510576.2 | XP_011508878.1 | 4 | 492 | |||
XM_011510577.2 | XP_011508879.1 | 4 | 679 | |||
XM_017003308.1 | XP_016858797.1 | 2 | 417 | |||
GenBank mRNA | AF411516.1 | - | 2 | 261 | ||
AF514993.1 | - | 2 | 218 | |||
AK289601.1 | - | 2 | 204 | |||
AK290760.1 | - | 2 | 173 | |||
AK298199.1 | - | 2 | 171 | |||
AK302350.1 | - | 3 | 333 | |||
BC035597.1 | - | 2 | 185 | |||
BC047029.1 | - | 2 | 248 | 5541023 | ||
BC061698.1 | - | 2 | 215 | 4132440 | ||
BI818393.1 | - | 2 | 240 | |||
BI821231.1 | - | 2 | 197 | |||
CX787176.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
