siRNA Details
Gene Symbol: ANKRD30BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027019.1 | 2 | 561 | |||
GenBank mRNA | BC057764.1 | - | 2 | 564 | ||
BC150521.1 | - | 2 | 192 | |||
BG719107.1 | - | 2 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
