siRNA Details
Gene Symbol: ANKRD30BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027019.1 | 5 | 1011 | |||
GenBank mRNA | BC057764.1 | - | 5 | 1014 | ||
BC150521.1 | - | 5 | 642 |
Gene Symbol: ANKRD30BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127901.1 | - | 2 | 1318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
