siRNA Details
Gene Symbol: DDX11L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045117.1 | 3 | 1521 | |||
GenBank mRNA | AM992876.1 | - | 3 | 1521 |
Gene Symbol: DDX11L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046018.2 | 3 | 1521 | |||
GenBank mRNA | AM992871.1 | - | 3 | 1521 |
Gene Symbol: DDX11L11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992877.1 | - | 3 | 1521 | ||
AM992881.1 | - | 3 | 1357 |
Gene Symbol: DDX11L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992878.1 | - | 3 | 1461 |
Gene Symbol: DDX11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024004.1 | 3 | 2023 | |||
NR_024005.2 | 3 | 1526 | ||||
GenBank mRNA | AK057285.1 | - | 2 | 2026 | ||
AK093685.1 | - | 2 | 2970 | |||
AK123946.1 | - | 3 | 2020 | |||
AM992870.1 | - | 3 | 1526 | |||
BC005070.1 | - | 1 | 598 |
Gene Symbol: DDX11L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992872.1 | - | 3 | 1521 |
Gene Symbol: DDX11L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992873.1 | - | 2 | 1164 |
Gene Symbol: DDX11L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034090.1 | 3 | 1521 | |||
NR_051985.1 | 3 | 1357 | ||||
GenBank mRNA | AM992875.1 | - | 3 | 1521 | ||
AM992880.1 | - | 3 | 1357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
