siRNA Details
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 10 | 2104 | ||
NM_001166276.1 | NP_001159748.1 | 9 | 2044 | |||
NM_001166277.1 | NP_001159749.1 | 9 | 1984 | |||
NM_014882.2 | NP_055697.1 | 9 | 2041 | |||
XM_005264675.2 | XP_005264732.1 | 10 | 1808 | |||
XM_005264676.2 | XP_005264733.1 | 9 | 1692 | |||
XM_011533206.2 | XP_011531508.1 | 10 | 1852 | |||
XM_011533207.2 | XP_011531509.1 | 10 | 1818 | |||
XM_011533208.2 | XP_011531510.1 | 11 | 1869 | |||
XM_011533209.2 | XP_011531511.1 | 7 | 1409 | |||
XM_011533210.2 | XP_011531512.1 | 5 | 936 | |||
XM_017005423.1 | XP_016860912.1 | 10 | 1712 | |||
XM_017005424.1 | XP_016860913.1 | 10 | 1709 | |||
XM_017005425.1 | XP_016860914.1 | 9 | 1595 | |||
XM_017005426.1 | XP_016860915.1 | 7 | 2046 | |||
XM_017005427.1 | XP_016860916.1 | 7 | 2043 | |||
GenBank mRNA | AK290396.1 | - | 10 | 1782 | ||
AK297056.1 | - | 9 | 1862 | |||
AK297157.1 | - | 10 | 1844 | |||
AK298669.1 | - | 9 | 1871 | |||
D29642.1 | - | 9 | 1854 | |||
EU794656.1 | - | 9 | 2041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
