siRNA Details
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004369.3 | NP_004360.2 | 8 | 3530 | ||
NM_057164.4 | NP_476505.3 | 6 | 2309 | |||
NM_057165.4 | NP_476506.3 | 7 | 2912 | |||
NM_057166.4 | NP_476507.3 | 5 | 1709 | |||
NM_057167.3 | NP_476508.2 | 7 | 2912 | |||
XM_005246065.1 | XP_005246122.1 | 7 | 2887 | |||
XM_005246066.1 | XP_005246123.1 | 5 | 2033 | |||
XM_006712253.1 | XP_006712316.1 | 8 | 3487 | |||
XM_011510574.1 | XP_011508876.1 | 8 | 3487 | |||
XM_017003303.1 | XP_016858792.1 | 13 | 3537 | |||
XM_017003304.1 | XP_016858793.1 | 10 | 1749 | |||
GenBank mRNA | AK092021.1 | - | 6 | 2235 | ||
AK303290.1 | - | 5 | 1676 | |||
AK304870.1 | - | 7 | 2847 | |||
BC144595.1 | - | 5 | 1650 | 9053125 | ||
BC150625.1 | - | 5 | 1650 | 9056995 | ||
BC171790.1 | - | 5 | 1650 | 9054444 | ||
BX647500.1 | - | 7 | 2874 | |||
X52022.1 | - | 8 | 3500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
