siRNA Details
Gene Symbol: FAM19A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005527.2 | NP_001005527.1 | 5 | 737 | ||
NM_182522.4 | NP_872328.1 | 5 | 819 | |||
XM_011533371.1 | XP_011531673.1 | 5 | 819 | |||
XM_011533372.1 | XP_011531674.1 | 5 | 695 | |||
GenBank mRNA | AK057890.1 | - | 4 | 435 | ||
AY325117.1 | - | 5 | 791 | |||
BC031566.1 | - | 5 | 651 | 5195201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
