siRNA Details
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 30 | 5289 | ||
NM_001304332.1 | NP_001291261.1 | 29 | 5262 | |||
XM_006711586.2 | XP_006711649.1 | 30 | 5180 | |||
XM_011510070.2 | XP_011508372.1 | 18 | 3209 | |||
XM_017002606.1 | XP_016858095.1 | 29 | 5153 | |||
XM_017002607.1 | XP_016858096.1 | 29 | 5045 | |||
XM_017002609.1 | XP_016858098.1 | 29 | 5184 | |||
GenBank mRNA | AK124535.1 | - | 5 | 3359 | ||
AK126624.1 | - | 17 | 3857 | |||
EF428334.1 | - | 35 | 5156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
