siRNA Details
Gene Symbol: CWC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020943.2 | NP_065994.1 | 15 | 1805 | ||
XM_005246726.2 | XP_005246783.1 | 15 | 1908 | |||
GenBank mRNA | AB046824.1 | - | 14 | 1624 | ||
AK025635.1 | - | 15 | 1662 | |||
AK026978.1 | - | 8 | 762 | |||
AK123647.1 | - | 2 | 762 | |||
BC016651.1 | - | 14 | 1629 | |||
BC031216.1 | - | 15 | 1805 | |||
BC041662.1 | - | 8 | 756 | |||
BC053573.1 | - | 12 | 1327 | |||
BC057826.1 | - | 15 | 1663 | |||
BC093952.1 | - | 14 | 1527 | 7939797 | ||
BC093954.1 | - | 14 | 1527 | 7939799 | ||
BC143433.1 | - | 13 | 1610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
