siRNA Details
Gene Symbol: CWC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020943.2 | NP_065994.1 | 20 | 3199 | ||
XM_005246726.2 | XP_005246783.1 | 20 | 3302 | |||
GenBank mRNA | AB046824.1 | - | 19 | 3018 | ||
AK026978.1 | - | 13 | 2156 | |||
AK123647.1 | - | 7 | 2156 | |||
BC041662.1 | - | 13 | 2150 | |||
BC053573.1 | - | 17 | 2721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
